siRNA Details
| Gene Symbol: Srpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016795.3 | NP_058075.2 | 8 | 644 | ||
| GenBank mRNA | AB012290.1 | - | 7 | 602 | ||
| AJ224115.1 | - | 8 | 638 | |||
| AK015754.1 | - | 5 | 386 | |||
| AK017187.1 | - | 8 | 641 | |||
| AK140761.1 | - | 54 | ||||
| AK141271.1 | - | 8 | 711 | |||
| AK151152.1 | - | 8 | 707 | |||
| AK161346.1 | - | 8 | 622 | |||
| AK161388.1 | - | 8 | 623 | |||
| BC005707.1 | - | 8 | 613 | 3157835 | ||
| BC050761.1 | - | 8 | 644 | 6771151 | ||

Srpk1
Genomic Map