siRNA Details
| Gene Symbol: Rab33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016858.2 | NP_058554.1 | 2 | 667 | ||
| GenBank mRNA | AB004665.1 | - | 2 | 617 | ||
| AB232629.1 | - | 2 | 301 | |||
| AK004974.1 | - | 2 | 403 | |||
| AK087967.1 | - | 2 | 407 | |||
| AK139821.1 | - | 2 | 653 | |||
| AK144706.1 | - | 2 | 406 | |||
| AK159674.1 | - | 2 | 402 | |||
| AK163783.1 | - | 2 | 654 | |||
| AK170483.1 | - | 2 | 296 | |||
| BC065076.1 | - | 2 | 667 | 6815992 | ||

Rab33b
Genomic Map