siRNA Details
| Gene Symbol: CIAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004804.2 | NP_004795.1 | 2, 3 | 489 | ||
| GenBank mRNA | AK223257.1 | - | 2, 3 | 410 | ||
| AK315568.1 | - | 2, 3 | 398 | |||
| BC001395.2 | - | 2, 3 | 353 | 3050587 | ||
| BC032812.1 | - | 2, 3 | 383 | 5219740 | ||
| BI460314.1 | - | 2, 3 | 497 | |||
| CR456802.1 | - | 2, 3 | 284 | |||
| EF011618.1 | - | 2, 3 | 383 | |||
| U63810.1 | - | 2, 3 | 440 | |||

CIAO1
Genomic Map