siRNA Details
| Gene Symbol: Stat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019963.1 | NP_064347.1 | 3, 4 | 434 | ||
| GenBank mRNA | AF187231.1 | - | 3, 4 | 428 | ||
| AF206162.1 | - | 3, 4 | 434 | |||
| AK149838.1 | - | 3, 4 | 431 | |||
| AK149924.1 | - | 3, 4 | 447 | |||
| AK150070.1 | - | 3, 4 | 446 | |||
| AK153405.1 | - | 1, 2 | 72 | |||

Stat2
Genomic Map