siRNA Details
| Gene Symbol: Tmsb4x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021278.2 | NP_067253.1 | 3 | 253 | ||
| GenBank mRNA | AK010659.1 | - | 3 | 197 | ||
| AK088783.1 | - | 3 | 197 | |||
| AK162286.1 | - | 3 | 198 | |||
| AK169108.1 | - | 3 | 200 | |||
| AK169718.1 | - | 3 | 197 | |||
| BC018286.1 | - | 3 | 169 | 3501378 | ||
| BQ127748.1 | - | 3 | 253 | |||
| X16053.1 | - | 3 | 282 | |||

Tmsb4x
Genomic Map