siRNA Details
| Gene Symbol: Stag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077712.2 | NP_001071180.1 | 8 | 1156 | ||
| NM_001290713.1 | NP_001277642.1 | 7 | 882 | |||
| NM_021465.4 | NP_067440.3 | 8 | 1016 | |||
| GenBank mRNA | AJ002636.1 | - | 7 | 858 | ||
| AK146718.1 | - | 8 | 1016 | |||
| AK168899.1 | - | 8 | 1016 | |||
| BC066041.1 | - | 8 | 897 | |||
| CO434048.1 | - | 6 | 703 | |||

Stag2
Genomic Map