siRNA Details
| Gene Symbol: Pln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141927.1 | NP_001135399.1 | 3 | 622 | ||
| NM_023129.5 | NP_075618.1 | 2 | 495 | |||
| GenBank mRNA | AK002622.1 | - | 2 | 475 | ||
| AK040718.1 | - | 2 | 466 | |||
| AK052199.1 | - | 2 | 429 | |||
| AK142708.1 | - | 2 | 450 | |||
| BB658675.1 | - | 2 | 495 | |||
| BC061097.1 | - | 2 | 488 | 6772294 | ||
| CA459471.1 | - | 3 | 615 | |||
| S46792.1 | - | 2 | 458 | |||

Pln
Genomic Map