siRNA Details
| Gene Symbol: Pln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141927.1 | NP_001135399.1 | 3 | 2410 | ||
| NM_023129.5 | NP_075618.1 | 2 | 2283 | |||
| GenBank mRNA | AF214653.1 | - | 1 | 1524 | ||
| AK002622.1 | - | 2 | 2263 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pln| Gene Symbol: Pln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141927.1 | NP_001135399.1 | 3 | 2410 | ||
| NM_023129.5 | NP_075618.1 | 2 | 2283 | |||
| GenBank mRNA | AF214653.1 | - | 1 | 1524 | ||
| AK002622.1 | - | 2 | 2263 | |||