siRNA Details
| Gene Symbol: Rce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023131.1 | NP_075620.1 | 8 | 999 | ||
| GenBank mRNA | AK012946.1 | - | 8 | 955 | ||
| AK209967.1 | - | 1 | 231 | |||
| BC004060.1 | - | 7 | 848 | 3590170 | ||
| BC048686.1 | - | 8 | 1048 | |||

Rce1
Genomic Map