siRNA Details
| Gene Symbol: Rad54b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039556.3 | NP_001034645.1 | 2, 3 | 253 | ||
| NM_001256145.1 | NP_001243074.1 | 2, 3 | 253 | |||
| GenBank mRNA | AK078439.1 | - | 2, 3 | 231 | ||
| BC057604.1 | - | 2, 3 | 201 | 6416213 | ||
| CJ160401.1 | - | 2, 3 | 254 | |||

Rad54b
Genomic Map