siRNA Details
| Gene Symbol: Stfa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025288.2 | NP_079564.1 | 3 | 338 | ||
| GenBank mRNA | AK014349.1 | - | 3 | 341 | ||
| AY169236.1 | - | 3 | 321 | |||
| BC107224.1 | - | 3 | 332 | 40051891 | ||
| BC107225.1 | - | 3 | 334 | 40051892 | ||
| M92419.1 | - | 3 | 322 | |||

Stfa3
Genomic Map