siRNA Details
| Gene Symbol: Rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029391.2 | NP_083667.1 | 6 | 652 | ||
| GenBank mRNA | AB232592.1 | - | 6 | 511 | ||
| AK005314.1 | - | 6 | 652 | |||
| AK140513.1 | - | 5 | 849 | |||
| AK170552.1 | - | 6 | 666 | |||
| AK219531.1 | - | 2 | 201 | |||
| BC007147.1 | - | 6 | 596 | 3600193 | ||
| BC096654.1 | - | 6 | 635 | 5056766 | ||

Rab4b
Genomic Map