siRNA Details
| Gene Symbol: Ptprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029928.2 | NP_084204.2 | 13 | 3410 | ||
| GenBank mRNA | AK147439.1 | - | 13 | 3412 | ||
| AK147668.1 | - | 13 | 3412 | |||
| AY077755.1 | - | 13 | 3288 | |||
| BC047086.1 | - | 3 | 470 | |||
| BC141006.1 | - | 13 | 3206 | 9055618 | ||
| BC145111.1 | - | 13 | 3206 | 9053659 | ||

Ptprb
Genomic Map