siRNA Details
| Gene Symbol: Slc2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031197.2 | NP_112474.2 | 3, 4 | 490 | ||
| GenBank mRNA | AK005068.1 | - | 3, 4 | 491 | ||
| AK165430.1 | - | 3, 4 | 425 | |||
| BC034675.1 | - | 3, 4 | 409 | 4236331 | ||
| X15684.1 | - | 3, 4 | 439 | |||
| X16986.1 | - | 3, 4 | 429 | |||

Slc2a2
Genomic Map