siRNA Details
| Gene Symbol: Sept8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252332.1 | NP_001239261.1 | 4 | 657 | ||
| NM_001252333.1 | NP_001239262.1 | 4 | 651 | |||
| NM_033144.2 | NP_149156.1 | 4 | 657 | |||
| GenBank mRNA | AB093215.1 | - | 4 | 507 | ||
| AF179996.1 | - | 3 | 353 | |||
| AK144178.1 | - | 4 | 493 | |||
| BC037707.1 | - | 4 | 445 | |||
| BC049819.1 | - | 4 | 472 | |||
| BC059248.1 | - | 4 | 593 | 6400429 | ||

Sept8
Genomic Map