siRNA Details
| Gene Symbol: Rheb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053075.3 | NP_444305.2 | 3 | 264 | ||
| GenBank mRNA | AB039919.1 | - | 3 | 314 | ||
| AK151213.1 | - | 3 | 296 | |||
| AK158213.1 | - | 336 | ||||
| AY197373.1 | - | 3 | 264 | |||
| BC012273.1 | - | 3 | 281 | 3710454 | ||

Rheb
Genomic Map