siRNA Details
| Gene Symbol: Hps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176785.3 | NP_789742.2 | 1 | 2596 | ||
| GenBank mRNA | AF536239.1 | - | 1 | 2541 | ||
| AK030582.1 | - | 1 | 2567 | |||
| AK040849.1 | - | 1 | 2543 | |||
| AK159588.1 | - | 1 | 2562 | |||
| AK172058.1 | - | 2 | 2347 | |||
| AK201433.1 | - | 1 | 114 | |||
| AK213448.1 | - | 1 | 113 | |||
| BC034055.1 | - | 1 | 2542 | 3673655 | ||

Hps6
Genomic Map