siRNA Details
| Gene Symbol: Rbbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009031.3 | NP_033057.3 | 2 | 410 | ||
| GenBank mRNA | AK076016.1 | - | 2 | 335 | ||
| AK078349.1 | - | 2 | 323 | |||
| AK082892.1 | - | 2 | 323 | |||
| AK132101.1 | - | 2 | 320 | |||
| AK135779.1 | - | 2 | 324 | |||
| AK135869.1 | - | 1 | 19 | |||
| AK135875.1 | - | 1 | 19 | |||
| AK135956.1 | - | 2 | 324 | |||
| AK136110.1 | - | 2 | 322 | |||
| AK145531.1 | - | 2 | 321 | |||
| AK145651.1 | - | 2 | 304 | |||
| AK146014.1 | - | 2 | 322 | |||
| AK146439.1 | - | 2 | 381 | |||
| AK146904.1 | - | 2 | 323 | |||
| AK146967.1 | - | 2 | 322 | |||
| AK147062.1 | - | 2 | 381 | |||
| AK148852.1 | - | 2 | 350 | |||
| AK153913.1 | - | 2 | 367 | |||
| AK160023.1 | - | 2 | 322 | |||
| BC003785.1 | - | 2 | 339 | 3600013 | ||
| U35142.1 | - | 2 | 349 | |||

Rbbp7
Genomic Map