siRNA Details
| Gene Symbol: Pde4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011056.3 | NP_035186.1 | 14 | 1676 | ||
| GenBank mRNA | AF536978.1 | - | 14 | 1676 | ||
| AK081466.1 | - | 7 | 1049 | |||
| AY388962.1 | - | 12 | 1289 | |||
| BC132107.1 | - | 13 | 1544 | 40130384 | ||
| BC132109.1 | - | 13 | 1544 | 40130386 | ||
| DQ665891.1 | - | 7 | 1146 | |||
| DQ665892.1 | - | 7 | 932 | |||
| DQ665893.1 | - | 8 | 1060 | |||
| DQ665894.1 | - | 8 | 904 | |||
| DQ665895.1 | - | 7 | 1187 | |||
| DQ665896.1 | - | 7 | 976 | |||
| EU489880.1 | - | 12 | 1532 | |||
| M94541.1 | - | 3 | 299 | |||

Pde4d
Genomic Map