siRNA Details
| Gene Symbol: Gin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026250.2 | NP_080526.2 | 7 | 1151 | ||
| GenBank mRNA | AK014287.1 | - | 7 | 1158 | ||
| AK015243.1 | - | 7 | 1130 | |||
| AK090002.1 | - | 7 | 869 | |||
| AK169361.1 | - | 7 | 1114 | |||
| BC033572.1 | - | 6 | 1096 | 5067692 | ||

Gin1
Genomic Map