siRNA Details
| Gene Symbol: Cdc42ep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026514.2 | NP_080790.1 | 2 | 1912 | ||
| GenBank mRNA | AK014281.1 | - | 2 | 1914 | ||
| AK019447.1 | - | 2 | 1914 | |||
| AK076782.1 | - | 2 | 1711 | |||
| AK150214.1 | - | 2 | 1879 | |||

Cdc42ep3
Genomic Map