siRNA Details
| Gene Symbol: 3110057O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031032.1 | - | 2436 | |||
| AK163334.1 | - | 5 | 1797 | |||
| BC051081.1 | - | 2686 | ||||
| BC145212.1 | - | 2123 | 9053763 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110057O12Rik| Gene Symbol: 3110057O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031032.1 | - | 2436 | |||
| AK163334.1 | - | 5 | 1797 | |||
| BC051081.1 | - | 2686 | ||||
| BC145212.1 | - | 2123 | 9053763 | |||