siRNA Details
| Gene Symbol: Frmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028127.3 | NP_082403.2 | 4 | 501 | ||
| GenBank mRNA | BC053929.1 | - | 4 | 501 | 6389695 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frmd6| Gene Symbol: Frmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028127.3 | NP_082403.2 | 4 | 501 | ||
| GenBank mRNA | BC053929.1 | - | 4 | 501 | 6389695 | |