siRNA Details
| Gene Symbol: Frmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028127.3 | NP_082403.2 | 13, 14 | 1821 | ||
| GenBank mRNA | AK029709.1 | - | 12, 13 | 1580 | ||
| AK203150.1 | - | 3, 4 | 262 | |||
| BC053929.1 | - | 13, 14 | 1821 | 6389695 | ||

Frmd6
Genomic Map