siRNA Details
| Gene Symbol: Wdr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172152.1 | NP_001165623.1 | 2, 3 | 218 | ||
| NM_028220.3 | NP_082496.3 | 3, 4 | 292 | |||
| GenBank mRNA | AK013889.1 | - | 2, 3 | 198 | ||
| AK020995.1 | - | 3, 4 | 315 | |||
| AK044241.1 | - | 2, 3 | 219 | |||
| BC028268.1 | - | 3, 4 | 303 | 5366098 | ||

Wdr17
Genomic Map