siRNA Details
| Gene Symbol: Scyl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286002.1 | NP_001272931.1 | 3, 4 | 645 | ||
| NM_001286003.1 | NP_001272932.1 | 3, 4 | 771 | |||
| NM_028776.5 | NP_083052.1 | 3, 4 | 832 | |||
| GenBank mRNA | AK004809.2 | - | 3, 4 | 572 | ||
| AK030824.1 | - | 3, 4 | 808 | |||
| AK044100.1 | - | 3, 4 | 732 | |||
| AK162937.1 | - | 3, 4 | 646 | |||
| BC043085.1 | - | 3, 4 | 533 | 6417440 | ||
| BC066800.1 | - | 3, 4 | 517 | |||

Scyl3
Genomic Map