siRNA Details
| Gene Symbol: Onecut3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139226.3 | NP_631972.2 | 2 | 1301 | ||
| GenBank mRNA | AY080897.1 | - | 2 | 1252 | ||
| BC058700.1 | - | 2 | 1301 | 6850438 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Onecut3| Gene Symbol: Onecut3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139226.3 | NP_631972.2 | 2 | 1301 | ||
| GenBank mRNA | AY080897.1 | - | 2 | 1252 | ||
| BC058700.1 | - | 2 | 1301 | 6850438 | ||