siRNA Details
| Gene Symbol: Rhbdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139228.3 | NP_631974.1 | 9 | 3706 | ||
| GenBank mRNA | AK041678.1 | - | 6 | 3284 | ||
| BC054784.1 | - | 9 | 3543 | 6817213 | ||
| BC056969.1 | - | 9 | 3543 | 6817213 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhbdl3| Gene Symbol: Rhbdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139228.3 | NP_631974.1 | 9 | 3706 | ||
| GenBank mRNA | AK041678.1 | - | 6 | 3284 | ||
| BC054784.1 | - | 9 | 3543 | 6817213 | ||
| BC056969.1 | - | 9 | 3543 | 6817213 | ||