siRNA Details
| Gene Symbol: Otoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139310.1 | NP_647471.1 | 15 | 1636 | ||
| GenBank mRNA | AK157904.1 | - | 15 | 1668 | ||
| AK158092.1 | - | 15 | 1751 | |||
| AK158280.1 | - | 5 | 609 | |||
| AK158326.1 | - | 5 | 605 | |||
| AY055122.1 | - | 15 | 1636 | |||

Otoa
Genomic Map