siRNA Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144931.3 | NP_659180.1 | 10 | 775 | ||
| GenBank mRNA | AK038756.1 | - | 5 | 1135 | ||
| AK146202.1 | - | 10 | 733 | |||
| AK162251.1 | - | 10 | 724 | |||
| AK166792.1 | - | 7 | 3650 | |||
| BC019163.1 | - | 10 | 755 | 3711115 | ||
| BC021510.1 | - | 10 | 756 | 5347277 | ||
| BC023680.1 | - | 10 | 757 | 5343341 | ||
| BC023897.1 | - | 10 | 732 | 5321187 | ||
| BC027124.1 | - | 10 | 719 | 5355631 | ||

Nae1
Genomic Map