siRNA Details
| Gene Symbol: Mbd3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144934.3 | NP_659183.3 | 3 | 497 | ||
| GenBank mRNA | AF503920.1 | - | 2 | 331 | ||
| AK135772.1 | - | 3 | 488 | |||
| BC064744.1 | - | 3 | 458 | 30051497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mbd3l2| Gene Symbol: Mbd3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144934.3 | NP_659183.3 | 3 | 497 | ||
| GenBank mRNA | AF503920.1 | - | 2 | 331 | ||
| AK135772.1 | - | 3 | 488 | |||
| BC064744.1 | - | 3 | 458 | 30051497 | ||