siRNA Details
| Gene Symbol: Lppr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290299.1 | NP_001277228.1 | 10 | 1920 | ||
| NM_144935.1 | NP_659184.1 | 9 | 1876 | |||
| GenBank mRNA | AK082453.1 | - | 10 | 1883 | ||
| AY438023.1 | - | 9 | 1876 | |||
| BC018242.1 | - | 9 | 1876 | 4503683 | ||
| BC023082.1 | - | 10 | 1897 | 5357160 | ||

Lppr2
Genomic Map