siRNA Details
| Gene Symbol: Tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144936.1 | NP_659185.1 | 6 | 1179 | ||
| GenBank mRNA | AK079262.1 | - | 6 | 1166 | ||
| AK144325.1 | - | 6 | 1404 | |||
| BC018222.1 | - | 6 | 1135 | 4221959 | ||
| BC026654.1 | - | 6 | 1179 | 4988382 | ||

Tmem45b
Genomic Map