siRNA Details
| Gene Symbol: Oas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145227.3 | NP_660262.2 | 5 | 1168 | ||
| GenBank mRNA | AB067535.1 | - | 5 | 1173 | ||
| AF418010.1 | - | 5 | 1108 | |||
| BC118538.1 | - | 5 | 1108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oas2| Gene Symbol: Oas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145227.3 | NP_660262.2 | 5 | 1168 | ||
| GenBank mRNA | AB067535.1 | - | 5 | 1173 | ||
| AF418010.1 | - | 5 | 1108 | |||
| BC118538.1 | - | 5 | 1108 | |||