siRNA Details
| Gene Symbol: BC017158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145590.2 | NP_663565.2 | 12 | 1297 | ||
| GenBank mRNA | AK148236.1 | - | 12 | 1288 | ||
| AK149645.1 | - | 12 | 1276 | |||
| AK154566.1 | - | 12 | 1265 | |||
| AK164488.1 | - | 12 | 1275 | |||
| AK165287.1 | - | 10 | 2740 | |||
| AK170611.1 | - | 11 | 1360 | |||
| BC017158.1 | - | 12 | 1279 | 4919869 | ||
| BC024080.1 | - | 11 | 907 | |||
| BC061499.1 | - | 2 | 1223 | |||
| BC094040.1 | - | 12 | 1281 | |||

BC017158
Genomic Map