siRNA Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293797.1 | NP_001280726.1 | 11 | 2511 | ||
| NM_145597.4 | NP_663572.1 | 12 | 2615 | |||
| GenBank mRNA | AK032711.1 | - | 8 | 2131 | ||
| AK049979.1 | - | 12 | 2614 | |||
| AK147962.1 | - | 12 | 2608 | |||

Tmem161a
Genomic Map