siRNA Details
| Gene Symbol: Tmem184c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145599.4 | NP_663574.3 | 4 | 791 | ||
| GenBank mRNA | AK033404.1 | - | 4 | 760 | ||
| AK164182.1 | - | 4 | 759 | |||
| BC004056.1 | - | 4 | 749 | 3589819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem184c| Gene Symbol: Tmem184c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145599.4 | NP_663574.3 | 4 | 791 | ||
| GenBank mRNA | AK033404.1 | - | 4 | 760 | ||
| AK164182.1 | - | 4 | 759 | |||
| BC004056.1 | - | 4 | 749 | 3589819 | ||