siRNA Details
Gene Symbol: Gm15455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161816.1 | NP_001155288.1 | 1 | 334 | ||
GenBank mRNA | AK139531.1 | - | 1083 | |||
BC157984.1 | - | 1 | 258 | |||
BC157993.1 | - | 1 | 258 |
Gene Symbol: Rbm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167775.1 | NP_001161247.1 | 3 | 804 | ||
NM_001167776.1 | NP_001161248.1 | 3 | 517 | |||
NM_145627.2 | NP_663602.1 | 3 | 517 | |||
GenBank mRNA | AK041871.1 | - | 3 | 492 | ||
AK089105.1 | - | 3 | 328 | |||
AK122208.1 | - | 3 | 435 | |||
AK145991.1 | - | 3 | 517 | |||
AK153736.1 | - | 3 | 805 | |||
AK167666.1 | - | 3 | 495 | |||
BC004674.1 | - | 2 | 202 | 3500403 | ||
BC093519.1 | - | 3 | 449 |
