siRNA Details
| Gene Symbol: Rbm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167775.1 | NP_001161247.1 | 11, 12 | 1858 | ||
| NM_001167776.1 | NP_001161248.1 | 10, 11 | 1343 | |||
| NM_145627.2 | NP_663602.1 | 11, 12 | 1574 | |||
| GenBank mRNA | AK041871.1 | - | 10, 11 | 1315 | ||
| AK089105.1 | - | 10, 11 | 1154 | |||
| AK122208.1 | - | 8, 9 | 3274 | |||
| AK145991.1 | - | 11, 12 | 1574 | |||
| AK153736.1 | - | 11, 12 | 1859 | |||
| AK167666.1 | - | 10, 11 | 1321 | |||
| BC004674.1 | - | 10, 11 | 1259 | 3500403 | ||
| BC093519.1 | - | 10, 11 | 1272 | |||

Rbm10
Genomic Map