siRNA Details
| Gene Symbol: Rbm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167775.1 | NP_001161247.1 | 9, 10 | 1614 | ||
| NM_001167776.1 | NP_001161248.1 | 8, 9 | 1096 | |||
| NM_145627.2 | NP_663602.1 | 9, 10 | 1327 | |||
| GenBank mRNA | AK041871.1 | - | 8, 9 | 1071 | ||
| AK089105.1 | - | 8, 9 | 907 | |||
| AK122208.1 | - | 7, 8 | 2856 | |||
| AK145991.1 | - | 9, 10 | 1327 | |||
| AK153736.1 | - | 9, 10 | 1615 | |||
| AK167666.1 | - | 8, 9 | 1074 | |||
| BC004674.1 | - | 8, 9 | 1012 | 3500403 | ||
| BC093519.1 | - | 8, 9 | 1028 | |||

Rbm10
Genomic Map