siRNA Details
Gene Symbol: Obox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145710.2 | NP_663756.2 | 2 | 501 | ||
GenBank mRNA | CN712286.1 | - | 2 | 501 | ||
CN721150.1 | - | 1 | 173 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Obox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145710.2 | NP_663756.2 | 2 | 501 | ||
GenBank mRNA | CN712286.1 | - | 2 | 501 | ||
CN721150.1 | - | 1 | 173 |