siRNA Details
| Gene Symbol: Obox6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145710.2 | NP_663756.2 | 2 | 508 | ||
| GenBank mRNA | CN712286.1 | - | 2 | 508 | ||
| CN721150.1 | - | 1 | 180 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obox6| Gene Symbol: Obox6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145710.2 | NP_663756.2 | 2 | 508 | ||
| GenBank mRNA | CN712286.1 | - | 2 | 508 | ||
| CN721150.1 | - | 1 | 180 | |||