siRNA Details
| Gene Symbol: Lctl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145835.2 | NP_665834.1 | 6 | 860 | ||
| GenBank mRNA | AF309072.1 | - | 6 | 861 | ||
| AK053515.1 | - | 6 | 749 | |||
| BC030631.1 | - | 2 | 185 | 1195880 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lctl| Gene Symbol: Lctl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145835.2 | NP_665834.1 | 6 | 860 | ||
| GenBank mRNA | AF309072.1 | - | 6 | 861 | ||
| AK053515.1 | - | 6 | 749 | |||
| BC030631.1 | - | 2 | 185 | 1195880 | ||