siRNA Details
| Gene Symbol: St8sia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145838.1 | NP_665837.1 | 7 | 767 | ||
| GenBank mRNA | AB059554.1 | - | 7 | 767 | ||
| AK085105.1 | - | 8 | 904 | |||
| AK156557.1 | - | 7 | 1304 | |||
| AK172400.1 | - | 7 | 832 | |||
| BC115912.1 | - | 7 | 722 | 40104779 | ||
| BC118063.1 | - | 7 | 722 | 40104780 | ||

St8sia6
Genomic Map