siRNA Details
| Gene Symbol: Vmn1r71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145848.3 | NP_665847.1 | 1 | 1160 | ||
| GenBank mRNA | AF394948.1 | - | 1 | 1158 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r71| Gene Symbol: Vmn1r71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145848.3 | NP_665847.1 | 1 | 1160 | ||
| GenBank mRNA | AF394948.1 | - | 1 | 1158 | ||