siRNA Details
| Gene Symbol: Armc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146205.2 | NP_666317.2 | 3 | 1642 | ||
| GenBank mRNA | AK030000.1 | - | 3 | 499 | ||
| AK131111.1 | - | 3 | 1642 | |||
| BC089363.1 | - | 3 | 883 | 5720275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc5| Gene Symbol: Armc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146205.2 | NP_666317.2 | 3 | 1642 | ||
| GenBank mRNA | AK030000.1 | - | 3 | 499 | ||
| AK131111.1 | - | 3 | 1642 | |||
| BC089363.1 | - | 3 | 883 | 5720275 | ||