siRNA Details
| Gene Symbol: Armc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146205.2 | NP_666317.2 | 6 | 3801 | ||
| GenBank mRNA | AK030000.1 | - | 6 | 2658 | ||
| AK131111.1 | - | 6 | 3801 | |||
| BC025196.1 | - | 3 | 1272 | |||
| BC032200.1 | - | 6 | 3146 | 5342166 | ||
| BC051105.1 | - | 2 | 1061 | |||
| BC089363.1 | - | 6 | 3042 | 5720275 | ||

Armc5
Genomic Map