siRNA Details
| Gene Symbol: Msantd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146222.2 | NP_666334.2 | 2 | 532 | ||
| GenBank mRNA | BC066002.1 | - | 2 | 532 | 5701381 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msantd2| Gene Symbol: Msantd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146222.2 | NP_666334.2 | 2 | 532 | ||
| GenBank mRNA | BC066002.1 | - | 2 | 532 | 5701381 | |