siRNA Details
| Gene Symbol: Hpdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146256.3 | NP_666368.1 | 1 | 1589 | ||
| GenBank mRNA | AK043902.1 | - | 1 | 1570 | ||
| AK049217.1 | - | 1 | 1569 | |||
| AK078929.1 | - | 1 | 1590 | |||
| BC034099.1 | - | 1 | 1404 | 4489103 | ||

Hpdl
Genomic Map