siRNA Details
| Gene Symbol: Fam199x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146261.2 | NP_666373.1 | 3, 4 | 851 | ||
| GenBank mRNA | AK039695.1 | - | 3, 4 | 779 | ||
| AK154624.1 | - | 3, 4 | 854 | |||
| BC031748.1 | - | 3, 4 | 816 | 3672969 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam199x| Gene Symbol: Fam199x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146261.2 | NP_666373.1 | 3, 4 | 851 | ||
| GenBank mRNA | AK039695.1 | - | 3, 4 | 779 | ||
| AK154624.1 | - | 3, 4 | 854 | |||
| BC031748.1 | - | 3, 4 | 816 | 3672969 | ||